Acyclovir 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,9-dihydro-2-amino-9-((2-hydroxyethoxy)methyl)-6h-purin-6-on, Acycloguanosine, 2-amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6h-purin-6-one, Acyclovir, 9-[(2-Hydroxyethoxy)methyl]guanine, 2-Amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6H-purin-6-one, 2-Amino-1,9-dihydro-9-[(2-hydroxyethoxy)methyl]-6H-purin-6-one, Aciclovir, 2-Amino-9-((2-hydroxyethoxy)methyl)-1H-purin-6(9H)-one
A118981-100G
A118981-100MG
A118981-25G
A118981-500G
A118981-50MG
76766-330EA
62.6
USD
76766-330
76766-332
76766-334
76766-336
77592-318
Acyclovir 98%
Acyclovir
Precaución: for Research Use Only
Formula:
C₈H₁₁N₅O₃ MW: 225.21 g/mol Boiling Pt: 595 °C (760 mm Hg) Melting Pt: 256…257 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00057880 CAS Number: 59277-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...