2-Amino-8-bromo[1,2,4]triazolo[1,5-a]pyridine 97%

Supplier: Ambeed
Warning

A193385-250MG A193385-1G A193385-10G
103816-530EA 26 USD
103816-530 103847-144 77029-900 77085-050 77667-608
2-Amino-8-bromo[1,2,4]triazolo[1,5-a]pyridine 97%
2-Amino-8-bromo[1,2,4]triazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₆H₅BrN₄
MW: 213.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658939
CAS Number: 1124382-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR