2-Amino-6-phenylpyridine 98%

Supplier: Ambeed
Warning

A180241-1G A180241-250MG
103807-526EA 42 USD
103807-526 103845-880 77083-366 77683-448
2-Amino-6-phenylpyridine 98%
2-Amino-6-phenylpyridine

Precaución: For research use only.

Formula: C₁₁H₁₀N₂
MW: 170.211 g/mol
Boiling Pt: 347.6 °C (760 mm Hg)
Melting Pt: 71…72 °C
Flash Pt: 191 °C
Storage Temperature: Ambient
MDL Number: MFCD00234719
CAS Number: 39774-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR