2-Amino-6-methylbenzothiazole 98%

Supplier: Ambeed
Warning

A420620-25G A420620-5G
103838-438EA 17 USD
103838-438 76842-062 77004-102
2-Amino-6-methylbenzothiazole 98%
2-Amino-6-methylbenzothiazole

Precaución: for Research Use Only

Formula: C₈H₈N₂S
MW: 164.23 g/mol
Boiling Pt: 322 °C (760 mm Hg)
Melting Pt: 140…142 °C
Flash Pt: 148.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00005789
CAS Number: 2536-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR