6-Methylanthranil acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-6-methylbenzoic acid

A256650-1G A256650-25G A256650-5G A256650-100G A256650-10G A256650-500G
103812-276EA 5 USD
103812-276 103852-140 77091-636 77091-638 77091-640 77250-046 77250-048 77250-050
6-Methylanthranil acid 97%
6-Methylanthranil acid

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 128…130 °C
Flash Pt: 144.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00007809
CAS Number: 4389-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR