2-Amino-6-methoxypyrazine 98%

Supplier: Ambeed
Warning

A208696-250MG A208696-1G A208696-5G A208696-100MG
103817-882EA 33 USD
103817-882 103848-568 76785-624 77086-892 77086-894 77667-754
2-Amino-6-methoxypyrazine 98%
2-Amino-6-methoxypyrazine

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MW: 125.13 g/mol
Boiling Pt: 266.1 °C (760 mm Hg)
Melting Pt: 112 °C
Flash Pt: 114.8 °C
Storage Temperature: Ambient
MDL Number: MFCD09909689
CAS Number: 6905-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR