6-Methoxyanthranilic acid 98%

Supplier: Ambeed
Warning

A288232-1G A288232-25G A288232-5G A288232-100G A288232-10G A288232-500G
103813-786EA 6 USD
103813-786 103834-928 76837-480 76837-482 76837-484 77262-766 77262-768 77262-770
6-Methoxyanthranilic acid 98%
6-Methoxyanthranilic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 341.5 °C (760 mm Hg)
Melting Pt: >85 °C
Flash Pt: 160.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01941328
CAS Number: 53600-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR