6-Fluoroanthranilic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-6-fluorobenzoic acid

A158617-100G A158617-10G A158617-500G A158617-25G
103843-356EA 5 USD
103843-356 76973-428 76973-430 76973-432 77319-218
6-Fluoroanthranilic acid 97%
6-Fluoroanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 308.8 °C (760 mm Hg)
Melting Pt: 170…172 °C
Flash Pt: 140.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00067781
CAS Number: 434-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR