2-Amino-6-cyanopyrazine 97%

Supplier: Ambeed
Warning

A175484-10G A175484-5G A175484-250MG
103807-044EA 94 USD
103807-044 103845-352 76784-420 77301-376 77871-878
2-Amino-6-cyanopyrazine 97%
2-Amino-6-cyanopyrazine

Precaución: for Research Use Only

Formula: C₅H₄N₄
Boiling Pt: 367.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD10697805
CAS Number: 59489-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR