2-Amino-6-chloropyrimidin-4(3H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-chloro-6-pyrimidinol, 2-Amino-4-chloro-6-hydroxypyrimidine

A271888-250MG A271888-1G A271888-25G A271888-100G
103813-072EA 7 USD
103813-072 103852-984 76787-202 77092-774 77092-776 77872-320
2-Amino-6-chloropyrimidin-4(3H)-one 97%
2-Amino-6-chloropyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₄H₄ClN₃O
Boiling Pt: 259.1 °C (760 mm Hg)
Melting Pt: 252 °C
Flash Pt: 110.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00006093
CAS Number: 1194-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR