2-Amino-6-chloropyrazine 98%

Supplier: Ambeed
Warning

A273183-100G A273183-10G A273183-500G A273183-5G A273183-25G A273183-1000G
103853-050EA 9 USD
103853-050 76816-246 76816-248 76816-250 77092-846 77305-124 77672-410
2-Amino-6-chloropyrazine 98%
2-Amino-6-chloropyrazine

Precaución: for Research Use Only

Formula: C₄H₄ClN₃
MW: 129.55 g/mol
Boiling Pt: 282.8 °C (760 mm Hg)
Melting Pt: 150…152 °C at N/A mmHg
Flash Pt: 124.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00055024
CAS Number: 33332-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR