2-Amino-6-chlorophenol 98%
Supplier: Ambeed
|
Warning
|
A158336-100MG
A158336-250MG
A158336-25G
A158336-1G
103805-120EA
14
USD
103805-120
103843-330
77028-490
77028-492
77300-322
77319-188
2-Amino-6-chlorophenol 98%
2-Amino-6-chlorophenol
Precaución: for Research Use Only
Formula:
C₆H₆ClNO MW: 143.5700 g/mol Boiling Pt: 247 °C (760 mm Hg) Melting Pt: 80…81 °C Flash Pt: 103 °C Storage Temperature: Ambient |
MDL Number:
MFCD07801210 CAS Number: 38191-33-2 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...