2-Amino-6-chlorobenzaldehyde 97%

Supplier: Ambeed
Warning

A303903-100MG A303903-250MG
103814-400EA 54 USD
103814-400 103835-566 77014-988 77014-990
2-Amino-6-chlorobenzaldehyde 97%
2-Amino-6-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆ClNO
Boiling Pt: 290.3±25.0 °C (760 mm Hg)
Flash Pt: 129.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10696880
CAS Number: 35490-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR