2-Amino-6-bromobenzothiazole 98%

Supplier: Ambeed
Warning

A176658-100G A176658-10G
103807-158EA 9 USD
103807-158 103845-484 77615-484 77615-486
2-Amino-6-bromobenzothiazole 98%
2-Amino-6-bromobenzothiazole

Precaución: Keep in a dark place, inert atmosphere, room temperature

Formula: C₇H₅BrN₂S
MW: 229.1 g/mol
Boiling Pt: 366.8 °C (760 mm Hg)
Melting Pt: 213…217 °C
Flash Pt: 175.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00152229
CAS Number: 15864-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR