6-Bromoanthranilic acid 98%

Supplier: Ambeed
Warning

A156961-100G A156961-10G A156961-25G A156961-1G A156961-5G
103804-944EA 6 USD
103804-944 103843-146 76997-572 76997-574 76997-576 77318-952 77318-954
6-Bromoanthranilic acid 98%
6-Bromoanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
Boiling Pt: 348.9 °C (760 mm Hg)
Flash Pt: 164.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03618455
CAS Number: 20776-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR