2-Amino-6-(trifluoromethyl)benzoic acid hydrochloride 97%

Supplier: Ambeed
Warning

A153275-5G A153275-250MG
103804-486EA 30 USD
103804-486 103842-658 77615-246 77748-052
2-Amino-6-(trifluoromethyl)benzoic acid hydrochloride 97%
2-Amino-6-(trifluoromethyl)benzoic acid hydrochloride

Precaución: Sealed in dry, at room temperature.

Formula: C₈H₇ClF₃NO₂
MW: 241.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30729973
CAS Number: 918667-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR