2-Amino-5-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

A181975-25G A181975-100G A181975-500G
103809-162EA 157 USD
103809-162 77083-574 77138-754 77138-756
2-Amino-5-nitrobenzonitrile 97%
2-Amino-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
Boiling Pt: 383.2 °C (760 mm Hg)
Melting Pt: 200…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00007362
CAS Number: 17420-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR