2-Amino-5-methylthiazole 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methylthiazol-2-ylamine

A126233-10G A126233-500G A126233-1000G A126233-25G A126233-100G
103808-328EA 146 USD
103808-328 77271-802 77271-804 77312-968 77312-970 77571-560
2-Amino-5-methylthiazole 97%
2-Amino-5-methylthiazole

Precaución: for Research Use Only

Formula: C₄H₆N₂S
MW: 114.17 g/mol
Boiling Pt: 232.5 °C (760 mm Hg)
Melting Pt: 93…98 °C
Flash Pt: 94.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00078317
CAS Number: 7305-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR