2-Amino-5-methyl-1,3,4-thiadiazole 98%

Supplier: Ambeed
Warning

A710744-1G A710744-25G A710744-100G A710744-10G A710744-500G A710744-5G
103840-694EA 6 USD
103840-694 76844-912 76844-914 76981-166 76981-168 76981-170 77308-180
2-Amino-5-methyl-1,3,4-thiadiazole 98%
2-Amino-5-methyl-1,3,4-thiadiazole

Precaución: For research use only

Formula: C₃H₅N₃S
MW: 115.16 g/mol
Melting Pt: 223…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00003110
CAS Number: 108-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR