2-Amino-5-methoxypyrimidine 97%

Supplier: Ambeed
Warning

A163780-100MG A163780-250MG A163780-1G
103805-766EA 19 USD
103805-766 103844-030 77028-718 77028-720 77320-142
2-Amino-5-methoxypyrimidine 97%
2-Amino-5-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MW: 125.131 g/mol
Boiling Pt: 313.1 °C (760 mm Hg)
Melting Pt: 86…88 °C
Flash Pt: 143 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00187864
CAS Number: 13418-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR