2-Amino-5-methoxybenzonitrile 97%

Supplier: Ambeed
Warning

A207360-1G A207360-5G A207360-10G A207360-25G
103817-788EA 16 USD
103817-788 103848-468 77086-744 77086-746 77248-988 77303-032
2-Amino-5-methoxybenzonitrile 97%
2-Amino-5-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂O
Boiling Pt: 302.2 °C (760 mm Hg)
Melting Pt: 46…47 °C
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD05149280
CAS Number: 23842-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR