5-Methoxyanthranilic acid 98%

Supplier: Ambeed
Warning

A430658-1G A430658-25G A430658-5G A430658-100G A430658-10G
103827-512EA 6 USD
103827-512 103838-578 76842-266 76842-268 76842-270 77252-460 77252-462
5-Methoxyanthranilic acid 98%
5-Methoxyanthranilic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00016509
CAS Number: 6705-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR