2-Amino-5-methoxybenzamide 98%

Supplier: Ambeed
Warning

A161599-10G A161599-250MG A161599-1G A161599-25G
103805-508EA 17 USD
103805-508 103843-746 76997-806 76997-808 77319-734 77671-286
2-Amino-5-methoxybenzamide 98%
2-Amino-5-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12674808
CAS Number: 1882-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR