2-Amino-5-iodopyrimidine 97%

Supplier: Ambeed
Warning

A203864-10G A203864-5G A203864-100G A203864-250MG
103817-500EA 9 USD
103817-500 103848-158 76814-386 77086-298 77883-473 77883-474
2-Amino-5-iodopyrimidine 97%
2-Amino-5-iodopyrimidine

Precaución: for Research Use Only

Formula: C₄H₄IN₃
Melting Pt: 218…223 °C
Flash Pt: 174.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01075666
CAS Number: 1445-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR