2-Amino-5-fluorobenzothiazole 98%

Supplier: Ambeed
Warning

A195513-250MG A195513-1G A195513-25G A195513-5G
103816-760EA 20 USD
103816-760 103847-372 77248-690 77302-468 77302-470 77302-472
2-Amino-5-fluorobenzothiazole 98%
2-Amino-5-fluorobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₅FN₂S
Boiling Pt: 312 °C (760 mm Hg)
Melting Pt: 181 °C
Flash Pt: 142.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05664560
CAS Number: 20358-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR