5-Fluoroanthranilic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-fluorobenzoic acid

A141157-10G A141157-500G A141157-25G A141157-5G A141157-100G
77272-472EA 15.45 USD
77272-472 77272-474 77316-100 77316-102 77592-678
5-Fluoroanthranilic acid 97%
5-Fluoroanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 318.2 °C (760 mm Hg)
Melting Pt: 181…183 °C
Flash Pt: 146.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00055566
CAS Number: 446-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR