2-Amino-5-ethylbenzoic acid 97%

Supplier: Ambeed
Warning

A152551-250MG A152551-1G
103804-402EA 173 USD
103804-402 103842-572 76871-662 77318-192
2-Amino-5-ethylbenzoic acid 97%
2-Amino-5-ethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD02358893
CAS Number: 2475-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR