2-Amino-5-cyano-N,3-dimethylbenzamide 95%

Supplier: Ambeed
Warning

A579604-1G A579604-100MG A579604-250MG A579604-25G A579604-5G
103828-856EA 18 USD
103828-856 76844-138 76878-800 76878-802 76878-804 76878-806
2-Amino-5-cyano-N,3-dimethylbenzamide 95%
2-Amino-5-cyano-N,3-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃O
MW: 189.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20690897
CAS Number: 890707-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR