2-Amino-5-chloropyridine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A215720-1G A215720-5G A215720-250MG
103818-512EA 17 USD
103818-512 103849-220 77087-768 77087-770 77249-214
2-Amino-5-chloropyridine-3-carboxylic acid 97%
2-Amino-5-chloropyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.5700 g/mol
Melting Pt: 262…266 °C
Storage Temperature: Ambient
MDL Number: MFCD09839254
CAS Number: 52833-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR