2-Amino-5-chloropyridine 98%

Supplier: Ambeed
Warning

A109539-1000G A109539-100G A109539-10G A109539-25G A109539-500G
103823-320EA 5 USD
103823-320 77270-762 77270-764 77270-766 77270-768 77270-770
2-Amino-5-chloropyridine 98%
2-Amino-5-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
Boiling Pt: 127…128 °C (760 mm Hg)
Melting Pt: 135…138 °C
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00006324
CAS Number: 1072-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR