2-Amino-5-chloropyridin-3-ol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-5-chloro-3-hydroxypyridine, 2-Amino-3-hydroxy-5-chloropyridine
A226667-1G
A226667-25G
A226667-5G
A226667-250MG
103819-258EA
6
USD
103819-258
103850-162
77088-898
77088-900
77303-774
77667-882
2-Amino-5-chloropyridin-3-ol 98%
2-Amino-5-chloropyridin-3-ol
Precaución: for Research Use Only
Formula:
C₅H₅ClN₂O MW: 144.5590 g/mol Melting Pt: 198…201 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD14706179 CAS Number: 40966-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...