2-Amino-5-chloronicotinonitrile 95%

Supplier: Ambeed
Warning

A167201-100MG A167201-250MG A167201-5G A167201-1G
103806-148EA 50 USD
103806-148 76824-348 76824-350 76824-352 77081-408
2-Amino-5-chloronicotinonitrile 95%
2-Amino-5-chloronicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16878647
CAS Number: 869557-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR