2-Amino-5-chloro-N,3-dimethylbenzamide 95%

Supplier: Ambeed
Warning

A121360-100G A121360-250MG A121360-25G A121360-5G A121360-1G
103800-064EA 11 USD
103800-064 76870-228 76870-230 76870-232 76870-234 76870-236 77297-828
2-Amino-5-chloro-N,3-dimethylbenzamide 95%
2-Amino-5-chloro-N,3-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD09030601
CAS Number: 890707-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR