5-Chloroanthranilic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-chlorobenzoic acid

A108243-100G A108243-10G A108243-25G A108243-500G A108243-5G
103807-994EA 97 USD
103807-994 103823-092 77135-798 77135-800 77135-802 77135-804 77283-698
5-Chloroanthranilic acid 97%
5-Chloroanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 204…206 °C
Flash Pt: 160.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00007838
CAS Number: 635-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR