2-Amino-5-chlorobenzamide 97%

Supplier: Ambeed
Warning

A387148-25G A387148-5G A387148-100G A387148-10G A387148-500G
103826-816EA 9 USD
103826-816 103837-858 76841-212 76841-214 77142-678 77142-680 77142-682
2-Amino-5-chlorobenzamide 97%
2-Amino-5-chlorobenzamide

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O
Boiling Pt: 264.1 °C (760 mm Hg)
Flash Pt: 113.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00017126
CAS Number: 5202-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR