2-Amino-5-chloro-3-iodopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-3-iodo-2-pyridinamine, 5-Chloro-3-iodo-2-pyridylamine, 5-Chloro-3-iodopyridin-2-amine

A208241-25G A208241-10G
103817-864EA 11 USD
103817-864 103848-552 77303-068 77683-592
2-Amino-5-chloro-3-iodopyridine 98%
2-Amino-5-chloro-3-iodopyridine

Precaución: For research use only.

Formula: C₅H₄ClIN₂
MW: 254.46 g/mol
Boiling Pt: 306.902 °C (760 mm Hg)
Melting Pt: 137…138 °C
Flash Pt: 139.41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07644578
CAS Number: 211308-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR