2-Amino-5-bromothiazole hydrobromide 98%

Supplier: Ambeed
Warning

A306717-25G A306717-5G A306717-100G A306717-1G A306717-500G
103835-652EA 6 USD
103835-652 76838-378 76838-380 76977-822 76977-824 76977-826
2-Amino-5-bromothiazole hydrobromide 98%
2-Amino-5-bromothiazole hydrobromide

Precaución: For research use only

Formula: C₃H₃BrN₂S
Boiling Pt: 192.5 °C (760 mm Hg)
Melting Pt: 165 °C
Flash Pt: 70.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00012712
CAS Number: 61296-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR