2-Amino-5-bromopyrazine 98%
Supplier: Ambeed
|
Warning
|
A170269-1G
A170269-500G
A170269-25G
A170269-5G
A170269-10G
A170269-100G
103844-736EA
10
USD
103844-736
76813-184
76813-186
77081-842
77081-844
77593-052
77611-656
2-Amino-5-bromopyrazine 98%
2-Amino-5-bromopyrazine
Precaución: for Research Use Only
Formula:
C₄H₄BrN₃ Boiling Pt: 274.2 °C (760 mm Hg) Melting Pt: 113…117 °C Flash Pt: 119.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00235015 CAS Number: 59489-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...