2-Amino-5-bromophenol 98%

Supplier: Ambeed
Warning

A205843-25G A205843-5G A205843-100G A205843-10G A205843-500G A205843-1G
103817-666EA 17 USD
103817-666 103848-332 77086-562 77086-564 77248-950 77248-952 77248-954 77302-968
2-Amino-5-bromophenol 98%
2-Amino-5-bromophenol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.0200 g/mol
Boiling Pt: 265 °C (760 mm Hg)
Flash Pt: 114.096 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095028
CAS Number: 38191-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR