2-Amino-5-bromonicotinonitrile 98%
Supplier: Ambeed
|
Warning
|
A232795-1G
A232795-25G
A232795-100MG
A232795-10G
A232795-250MG
A232795-5G
103810-852EA
9
USD
103810-852
103850-604
77089-536
77089-538
77165-576
77165-578
77165-580
77303-944
2-Amino-5-bromonicotinonitrile 98%
2-Amino-5-bromonicotinonitrile
Precaución: for Research Use Only
Formula:
C₆H₄BrN₃ Flash Pt: 126.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD08688591 CAS Number: 709652-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...