2-Amino-5-bromonicotinamide 95%

Supplier: Ambeed
Warning

A112119-100MG A112119-250MG
103797-954EA 132 USD
103797-954 103823-788 77025-810 77025-812
2-Amino-5-bromonicotinamide 95%
2-Amino-5-bromonicotinamide

Precaución: for Research Use Only

Formula: C₆H₆BrN₃O
MW: 216.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16688836
CAS Number: 58483-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR