2-Amino-5-bromobenzothiazole 97%

Supplier: Ambeed
Warning

A129033-100G A129033-10G A129033-250MG A129033-25G A129033-1G A129033-5G
103801-218EA 10 USD
103801-218 77109-102 77109-104 77109-106 77109-108 77313-576 77313-578
2-Amino-5-bromobenzothiazole 97%
2-Amino-5-bromobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₅BrN₂S
MW: 229.1 g/mol
Boiling Pt: 366.8 °C (760 mm Hg)
Melting Pt: 194…198 °C
Flash Pt: 175.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00577806
CAS Number: 20358-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR