2-Amino-5-bromobenzonitrile 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-2-cyanoaniline
A147924-100G
A147924-10G
A147924-25G
A147924-5G
103841-964EA
6
USD
103841-964
77137-648
77137-650
77317-380
77317-382
2-Amino-5-bromobenzonitrile 95%
2-Amino-5-bromobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₅BrN₂ Boiling Pt: 288.4 °C (760 mm Hg) Melting Pt: 96…100 °C Flash Pt: 128.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00158946 CAS Number: 39263-32-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...