5-Bromoanthranilic acid 98%

Supplier: Ambeed
Warning

A184792-25G A184792-100G A184792-500G
103809-188EA 148 USD
103809-188 103846-350 77083-974 77138-848 77138-850
5-Bromoanthranilic acid 98%
5-Bromoanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
Boiling Pt: 342.4 °C (760 mm Hg)
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00007823
CAS Number: 5794-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR