2-Amino-5-bromobenzaldehyde 98%

Supplier: Ambeed
Warning

A131124-10G A131124-25G A131124-250MG A131124-100G
103801-526EA 15 USD
103801-526 77027-070 77314-006 77599-890 77881-041
2-Amino-5-bromobenzaldehyde 98%
2-Amino-5-bromobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆BrNO
Boiling Pt: 290.2 °C (760 mm Hg)
Melting Pt: 75…77 °C
Flash Pt: 129.349 °C
Storage Temperature: Freezer
MDL Number: MFCD10696879
CAS Number: 29124-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR