2-Amino-5-bromo-6-chloropyrazine 98%

Supplier: Ambeed
Warning

A900854-100G A900854-10G A900854-25G A900854-1G A900854-500G
103841-350EA 9 USD
103841-350 76792-074 76792-076 76845-620 77308-496 77603-250
2-Amino-5-bromo-6-chloropyrazine 98%
2-Amino-5-bromo-6-chloropyrazine

Precaución: for Research Use Only

Formula: C₄H₃BrClN₃
MW: 208.45 g/mol
Boiling Pt: 311.8 °C (760 mm Hg)
Flash Pt: 142.4 °C
Storage Temperature: Ambient
MDL Number: MFCD09909642
CAS Number: 173253-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR