2-Amino-5-bromo-4-methoxypyrimidine 95%

Supplier: Ambeed
Warning

A211070-1G A211070-5G
103818-076EA 21 USD
103818-076 103848-770 77303-172 77303-174
2-Amino-5-bromo-4-methoxypyrimidine 95%
2-Amino-5-bromo-4-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃O
Melting Pt: 118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08460376
CAS Number: 36082-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR