2-Amino-5-bromo-4-chloro-3-nitropyridine 97%

Supplier: Ambeed
Warning

A117475-250MG A117475-25G A117475-1G A117475-5G
103798-896EA 37 USD
103798-896 103824-784 77245-966 77245-968 77311-564 77311-566
2-Amino-5-bromo-4-chloro-3-nitropyridine 97%
2-Amino-5-bromo-4-chloro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrClN₃O₂
MW: 252.4500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11110693
CAS Number: 942947-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR