2-Amino-5-bromo-3-pyridinol 98%

Supplier: Ambeed
Warning

A153764-100G A153764-10G A153764-250MG A153764-1G A153764-25G A153764-5G
103804-560EA 6 USD
103804-560 103842-738 76783-506 76783-508 76783-510 77318-418 77318-420 77318-422
2-Amino-5-bromo-3-pyridinol 98%
2-Amino-5-bromo-3-pyridinol

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
Boiling Pt: 295.1 °C (760 mm Hg)
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD09744143
CAS Number: 39903-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR