2-Amino-5-bromo-3-methylpyrazine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-methylpyrazin-2-amine

A163949-100MG A163949-1G A163949-5G A163949-250MG
103805-794EA 58 USD
103805-794 103844-060 76783-986 77320-182 77320-184 77667-292
2-Amino-5-bromo-3-methylpyrazine 98%
2-Amino-5-bromo-3-methylpyrazine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃
MW: 188.0300 g/mol
Boiling Pt: 272.4 °C (760 mm Hg)
Flash Pt: 118.532 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08705773
CAS Number: 74290-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR